(R)-N-((S)-2-(Diphenylphosphanyl)-1-(2-(diphenylphosphanyl)-4,5-dimethoxyphenyl)ethyl)-2-methylpropane-2-sulfinamide ≥95%

Supplier: Ambeed
Warning

A1487058-100MG A1487058-50MG
77597-600EA 401.8 USD
77597-600 77624-428
(R)-N-((S)-2-(Diphenylphosphanyl)-1-(2-(diphenylphosphanyl)-4,5-dimethoxyphenyl)ethyl)-2-methylpropane-2-sulfinamide ≥95%
(R)-N-((S)-2-(Diphenylphosphanyl)-1-(2-(diphenylphosphanyl)-4,5-dimethoxyphenyl)ethyl)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₃₈H₄₁NO₃P₂S
MW: 653.75 g/mol
MDL Number: MFCD32697182
CAS Number: 2565792-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR