(2-(2,2,2-Trifluoroethyl)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1486552-100MG A1486552-250MG
77624-424EA 244.11 USD
77624-424 77624-426
(2-(2,2,2-Trifluoroethyl)phenyl)boronic acid ≥95%
(2-(2,2,2-Trifluoroethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂
MW: 203.95 g/mol
MDL Number: MFCD22548091
CAS Number: 2408428-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR