(2R,3R,4R,5R,6R)-5-Acetamido-2-(acetoxymethyl)-6-(3-(((benzyloxy)carbonyl)amino)propoxy)tetrahydro-2H-pyran-3,4-diyl diacetate ≥98%

Supplier: Ambeed
Warning

A1486439-250MG A1486439-5G
77875-354EA 11 USD
77875-354 77875-355
(2R,3R,4R,5R,6R)-5-Acetamido-2-(acetoxymethyl)-6-(3-(((benzyloxy)carbonyl)amino)propoxy)tetrahydro-2H-pyran-3,4-diyl diacetate ≥98%
(2R,3R,4R,5R,6R)-5-Acetamido-2-(acetoxymethyl)-6-(3-(((benzyloxy)carbonyl)amino)propoxy)tetrahydro-2H-pyran-3,4-diyl diacetate

Precaución: for Research Use Only

Formula: C₂₅H₃₄N₂O₁₁
CAS Number: 143918-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR