1-Methoxynaphthalene-2-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (1-Methoxynaphthalen-2-yl)boronic acid

A1486417-100MG A1486417-1G A1486417-250MG A1486417-5G
77875-350EA 8 USD
77875-350 77875-351 77875-352 77875-353
1-Methoxynaphthalene-2-boronic acid ≥98%
1-Methoxynaphthalene-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BO₃
MDL Number: MFCD18412861
CAS Number: 252670-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR