(5-(1-Cyanocyclopropyl)-2-fluorophenyl)boronic acid ≥98%

Supplier: Ambeed

A148611-1G A148611-250MG A148611-5G
77885-231EA 189 USD
77885-231 77885-232 77885-233
(5-(1-Cyanocyclopropyl)-2-fluorophenyl)boronic acid ≥98%
(5-(1-Cyanocyclopropyl)-2-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BFNO₂
MDL Number: MFCD17926483
CAS Number: 1256345-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR