4-Hydroxy-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione ≥97%

Supplier: Ambeed
Warning

A1486108-250MG A1486108-1G
77624-408EA 200.85 USD
77624-408 77875-331
4-Hydroxy-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione ≥97%
4-Hydroxy-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₅
CAS Number: 2244568-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR