α,α,α-Trifluoro-2-iodotoluene 98+% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A1485733-25G A1485733-100G A1485733-5G
77597-580EA 13.33 USD
77597-580 77875-329 77875-330
α,α,α-Trifluoro-2-iodotoluene 98+% stabilized
α,α,α-Trifluoro-2-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₄F₃I
MDL Number: MFCD00001041
CAS Number: 444-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR