(9H-Fluoren-9-yl)methyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-DOOA HCl

A1483994-1G A1483994-250MG A1483994-25G A1483994-5G
77597-576EA 98.4 USD
77597-576 77597-578 77624-394 77875-328
(9H-Fluoren-9-yl)methyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate hydrochloride ≥95%
(9H-Fluoren-9-yl)methyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₇ClN₂O₄
MW: 406.9031 g/mol
MDL Number: MFCD04112696
CAS Number: 868599-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR