1-Boc-4-(2-chloroethyl)piperazine ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Chloroethyl)piperazine,N1-BOC protected, tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate

A1483780-5G A1483780-1G A1483780-25G
77592-796EA 33.83 USD
77592-796 77597-572 77624-384
1-Boc-4-(2-chloroethyl)piperazine ≥98%
1-Boc-4-(2-chloroethyl)piperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₁ClN₂O₂
MW: 248.751 g/mol
MDL Number: MFCD08443993
CAS Number: 208167-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR