Methoxypolyethylene glycol amine, M.W. ~550

Supplier: Ambeed
Warning

Synonyms: Amino polyethylene glycol monomethyl ether-average M.W. 2000, m-PEG12-amine, mPEG-amine, Amino polyethylene glycol monomethyl ether, mPEG-NH2

A1483518-100MG A1483518-250MG A1483518-1G
77597-566EA 28.7 USD
77597-566 77597-568 77624-376
Methoxypolyethylene glycol amine, M.W. ~550
Methoxypolyethylene glycol amine

Precaución: for Research Use Only

Formula: H₂NCH₂CH₂(OCH₂CH₂)nOCH₃
MDL Number: MFCD00131599
CAS Number: 80506-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR