m-PEG-thiol, M.W. ~5000 ≥97%

Supplier: Ambeed
Danger

A1483154-5G A1483154-250MG
77624-368EA 143.17 USD
77624-368 77875-310
m-PEG-thiol, M.W. ~5000 ≥97%
m-PEG-thiol

Precaución: for Research Use Only

MDL Number: MFCD21363241
CAS Number: 134874-49-0
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR