Benzo[d]oxazole-5-carbaldehyde ≥96%
Supplier: Ambeed
|
Warning
|
A148278-100MG
A148278-1G
A148278-250MG
77683-260EA
55
USD
77683-260
77683-261
77683-262
Benzo[d]oxazole-5-carbaldehyde ≥96%
Benzo[d]oxazole-5-carbaldehyde
Precaución: For research use only.
Formula:
C₈H₅NO₂ MW: 147.13 g/mol |
MDL Number:
MFCD20527251 CAS Number: 638192-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...