2-(3,5-Bis(trifluoromethyl)phenyl)-9-bromo-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate ≥97%

Supplier: Ambeed
Warning

A1482779-100MG A1482779-250MG
77592-788EA 200.9 USD
77592-788 77597-556
2-(3,5-Bis(trifluoromethyl)phenyl)-9-bromo-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate ≥97%
2-(3,5-Bis(trifluoromethyl)phenyl)-9-bromo-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₀H₁₃BBrF₁₀N₃O
MW: 592.04 g/mol
CAS Number: 2408127-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR