4-((2-Aminoethyl)amino)butan-1-ol dihydrochloride ≥97%

Supplier: Ambeed
Warning

A1482647-100MG A1482647-250MG
77875-291EA 66 USD
77875-291 77875-292
4-((2-Aminoethyl)amino)butan-1-ol dihydrochloride ≥97%
4-((2-Aminoethyl)amino)butan-1-ol dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₈Cl₂N₂O
MW: 205.1259 g/mol
MDL Number: MFCD34476164
CAS Number: 2757729-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR