Boc-3,5-diiodo-L-tyrosine 98+%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid, Boc-Tyr(3,5-DiI)-OH, Boc-3,5-diiodo-Tyr-OH

A1482482-25G A1482482-5G
77875-283EA 50 USD
77875-283 77875-284
Boc-3,5-diiodo-L-tyrosine 98+%
Boc-3,5-diiodo-L-tyrosine

Precaución: for Research Use Only

Formula: C₁₄H₁₇I₂NO₅
MW: 533.101 g/mol
MDL Number: MFCD00058515
CAS Number: 62129-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR