N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol 98+%

Supplier: Ambeed
Warning

Synonyms: 2-(tert-Butoxycarbonylamino)-2-phenylethanol, N-Boc-DL-2-phenylglycinol, Boc-DL-Phenylglycinol, tert-Butyl (2-hydroxy-1-phenylethyl)carbamate

A1482474-5G A1482474-100G
77597-546EA 8.2 USD
77597-546 77875-282
N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol 98+%
N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
MW: 237.30 g/mol
MDL Number: MFCD01883039
CAS Number: 67341-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR