O-Benzyl-N-benzyloxycarbonyl-L-threonine ≥98%

Supplier: Ambeed
Warning

Synonyms: Z-Thr(Bzl)-OH, Cbz-Thr(Bzl)-OH, (2S,3R)-3-(Benzyloxy)-2-(((benzyloxy)carbonyl)amino)butanoic acid

A1482473-1G A1482473-25G A1482473-5G
77597-540EA 17.43 USD
77597-540 77597-542 77597-544
O-Benzyl-N-benzyloxycarbonyl-L-threonine ≥98%
O-Benzyl-N-benzyloxycarbonyl-L-threonine

Precaución: for Research Use Only

Formula: C₁₉H₂₁NO₅
MW: 343.379 g/mol
MDL Number: MFCD00077042
CAS Number: 69863-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR