Nɑ-Carbobenzoxy-DL-asparagine 98+%

Supplier: Ambeed
Warning

Synonyms: Nɑ-Cbz-DL-asparagine, Z-DL-Asn-OH, N-Benzyloxycarbonyl-DL-asparagine, Cbz-DL-Asn-OH, 4-Amino-2-(Cbz-amino)-4-oxobutyric Acid

A1482464-5G A1482464-25G
77597-538EA 18.45 USD
77597-538 77875-276
Nɑ-Carbobenzoxy-DL-asparagine 98+%
Nalpha-Carbobenzoxy-DL-asparagine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₅
MW: 266.25 g/mol
MDL Number: MFCD00065101
CAS Number: 29880-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR