(4-Chloropyridin-2-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A148240-100MG A148240-1G A148240-250MG
77883-172EA 125 USD
77883-172 77883-173 77883-174
(4-Chloropyridin-2-yl)boronic acid ≥95%
(4-Chloropyridin-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₅BClNO₂
MDL Number: MFCD12401036
CAS Number: 870459-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR