tert-Butyl hept-6-yn-1-ylcarbamate ≥95%

Supplier: Ambeed
Warning

A1482128-1G A1482128-250MG A1482128-5G
77597-532EA 214.22 USD
77597-532 77597-534 77875-264
tert-Butyl hept-6-yn-1-ylcarbamate ≥95%
tert-Butyl hept-6-yn-1-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₂
MDL Number: MFCD24467680
CAS Number: 1192373-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR