tert-Butyl 3,6,9,12-tetraoxapentadec-14-yn-1-ylcarbamate ≥95%

Supplier: Ambeed
Warning

Synonyms: Boc-N-Amido-PEG4-propargyl

A1482027-100MG A1482027-250MG A1482027-1G
77597-528EA 33.83 USD
77597-528 77597-530 77624-358
tert-Butyl 3,6,9,12-tetraoxapentadec-14-yn-1-ylcarbamate ≥95%
tert-Butyl 3,6,9,12-tetraoxapentadec-14-yn-1-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₆H₂₉NO₆
MDL Number: MFCD30182022
CAS Number: 1219810-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR