3-(2-Bromoethoxy)prop-1-yne ≥95%

Supplier: Ambeed
Danger

Synonyms: 2-Bromoethyl(2-propynyl) ether

A1481739-1G A1481739-250MG A1481739-5G
77597-520EA 209.1 USD
77597-520 77624-350 77624-352
3-(2-Bromoethoxy)prop-1-yne ≥95%
3-(2-Bromoethoxy)prop-1-yne

Precaución: for Research Use Only

Formula: C₅H₇BrO
MW: 163.01 g/mol
MDL Number: MFCD00099299
CAS Number: 18668-74-1
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR