2,4-Butanesultone ≥98%

Supplier: Ambeed
Danger

A148134-250MG A148134-5G
77881-155EA 13 USD
77881-155 77881-156
2,4-Butanesultone ≥98%
2,4-Butanesultone

Precaución: for Research Use Only

Formula: C₄H₈O₃S
MDL Number: MFCD01941639
CAS Number: 1121-03-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR