2-Fluoro-6-methoxy-3-methylphenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-6-methoxy-3-methylphenyl)boronic acid

A148022-100MG A148022-250MG A148022-5G
77883-167EA 24 USD
77883-167 77883-168 77883-169
2-Fluoro-6-methoxy-3-methylphenylboronic acid ≥98%
2-Fluoro-6-methoxy-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MDL Number: MFCD11856029
CAS Number: 1451392-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR