(S)-1-(3-Bromo-4-fluorophenyl)ethanamine ≥95%

Supplier: Ambeed
Warning

A1480142-100MG A1480142-250MG
77875-245EA 32 USD
77875-245 77875-246
(S)-1-(3-Bromo-4-fluorophenyl)ethanamine ≥95%
(S)-1-(3-Bromo-4-fluorophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₉BrFN
MDL Number: MFCD06761803
CAS Number: 1212376-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR