3-Boc-aminoethylazetidine ≥96%

Supplier: Ambeed
Warning

Synonyms: 3-[2-(Boc-amino)ethyl]azetidine, 3-(N-BOC)-(2-aminoethyl)azetidine, 3-Azetidinecarboxylic acid, 1-(2-aminoethyl)-, 1,1-dimethylethyl ester, 3-Boc-Aminoethylazetidine, 2-Methyl-2-propanyl 1-(2-aminoethyl)-3-azetidinecarboxylate

A147898-250MG A147898-100MG
77611-366EA 165.83 USD
77611-366 77685-242
3-Boc-aminoethylazetidine ≥96%
3-Boc-aminoethylazetidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.282 g/mol
MDL Number: MFCD09878605
CAS Number: 162696-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR