2-Cyano-6-fluorophenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (2-Cyano-6-fluorophenyl)boronic acid

A1477088-250MG A1477088-1G
77624-334EA 20.6 USD
77624-334 77875-222
2-Cyano-6-fluorophenylboronic acid ≥98%
2-Cyano-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BFNO₂
MDL Number: MFCD09033623
CAS Number: 656235-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR