Sodium thiocyanate ≥98%

Supplier: Ambeed
Danger

A1476660-1000G A1476660-100G A1476660-500G
77597-502EA 11.28 USD
77597-502 77597-504 77597-506
Sodium thiocyanate ≥98%
Sodium thiocyanate

Precaución: for Research Use Only

Formula: NaSCN
MDL Number: MFCD00011123
CAS Number: 540-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR