5-(Hydroxymethyl)-2-nitrobenzonitrile 95+%

Supplier: Ambeed
Warning

A1476212-100MG A1476212-1G A1476212-250MG
77875-203EA 34 USD
77875-203 77875-204 77875-205
5-(Hydroxymethyl)-2-nitrobenzonitrile 95+%
5-(Hydroxymethyl)-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
CAS Number: 2757731-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR