(S)-7-(4-(1-(Methylsulfonyl)piperidin-4-yl)phenyl)-N-(morpholin-2-ylmethyl)pyrido[3,4-b]pyrazin-5-amine ≥95%

Supplier: Ambeed
Warning

Synonyms: Sovleplenib

A1476009-10MG A1476009-5MG A1476009-1MG A1476009-100MG A1476009-1G A1476009-50MG A1476009-250MG
77600-104EA 45.1 USD
77600-104 77600-106 77597-492 77624-316 77624-318 77624-320 77875-198
(S)-7-(4-(1-(Methylsulfonyl)piperidin-4-yl)phenyl)-N-(morpholin-2-ylmethyl)pyrido[3,4-b]pyrazin-5-amine ≥95%
(S)-7-(4-(1-(Methylsulfonyl)piperidin-4-yl)phenyl)-N-(morpholin-2-ylmethyl)pyrido[3,4-b]pyrazin-5-amine

Precaución: for Research Use Only

Formula: C₂₄H₃₀N₆O₃S
MW: 482.6 g/mol
CAS Number: 1415792-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR