1,1'-[(5R)-2,2',3,3'-Tetrahydro[5,5'-bi-1,4-benzodioxin]-6,6'-diyl]bis[1,1-bis[3,5-bis(trifluoromethyl)phenyl]phosphine] ≥97%

Supplier: Ambeed
Warning

A1475929-250MG A1475929-1G
77597-482EA 277.78 USD
77597-482 77624-314
1,1'-[(5R)-2,2',3,3'-Tetrahydro[5,5'-bi-1,4-benzodioxin]-6,6'-diyl]bis[1,1-bis[3,5-bis(trifluoromethyl)phenyl]phosphine] ≥97%
1,1'-[(5R)-2,2',3,3'-Tetrahydro[5,5'-bi-1,4-benzodioxin]-6,6'-diyl]bis[1,1-bis[3,5-bis(trifluoromethyl)phenyl]phosphine]

Precaución: for Research Use Only

Formula: C₄₈H₂₄F₂₄O₄P₂
MW: 1182.6100 g/mol
CAS Number: 1306747-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR