(S)-2,6-Bis(3,5-di-tert-butyl-4-methoxyphenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%

Supplier: Ambeed
Warning

A1475509-100MG A1475509-250MG A1475509-1G
77624-304EA 208.06 USD
77624-304 77624-306 77875-186
(S)-2,6-Bis(3,5-di-tert-butyl-4-methoxyphenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%
(S)-2,6-Bis(3,5-di-tert-butyl-4-methoxyphenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₅₀H₆₅O₆P
CAS Number: 2757287-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR