Ethyl 2,2-difluoro-5-oxohexahydro-1H-pyrrolizine-7a-carboxylate ≥97%

Supplier: Ambeed
Warning

A1475216-250MG A1475216-100MG
77592-766EA 112.75 USD
77592-766 77597-470
Ethyl 2,2-difluoro-5-oxohexahydro-1H-pyrrolizine-7a-carboxylate ≥97%
Ethyl 2,2-difluoro-5-oxohexahydro-1H-pyrrolizine-7a-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₃F₂NO₃
MW: 233.21 g/mol
MDL Number: MFCD31926295
CAS Number: 1788873-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR