tert-Butyl (32-bromo-3,6,9,12,15,18,21,24,27,30-decaoxadotriacontyl)carbamate ≥95%

Supplier: Ambeed
Warning

Synonyms: t-Boc-N-amido-PEG10-Br

A1475171-100MG A1475171-50MG
77597-466EA 224.48 USD
77597-466 77597-468
tert-Butyl (32-bromo-3,6,9,12,15,18,21,24,27,30-decaoxadotriacontyl)carbamate ≥95%
tert-Butyl (32-bromo-3,6,9,12,15,18,21,24,27,30-decaoxadotriacontyl)carbamate

Precaución: for Research Use Only

Formula: C₂₇H₅₄BrNO₁₂
CAS Number: 2787474-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR