(S)-4-(tert-Butyl)-2-(2-(diphenylphosphino)benzyl)-4,5-dihydrooxazole ≥97%

Supplier: Ambeed
Warning

A1475105-100MG A1475105-1G A1475105-250MG A1475105-5G
77875-142EA 155 USD
77875-142 77875-143 77875-144 77875-145
(S)-4-(tert-Butyl)-2-(2-(diphenylphosphino)benzyl)-4,5-dihydrooxazole ≥97%
(S)-4-(tert-Butyl)-2-(2-(diphenylphosphino)benzyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₆H₂₈NOP
CAS Number: 761402-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR