4-(4-Fluoro-3-(2-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[1,5-a]pyrazine-7-carbonyl)benzyl)phthalazin-1(2H)-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Fluzoparib

A1474773-10MG A1474773-1MG A1474773-5MG
77597-448EA 51.25 USD
77597-448 77597-450 77597-452
4-(4-Fluoro-3-(2-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[1,5-a]pyrazine-7-carbonyl)benzyl)phthalazin-1(2H)-one ≥98%
4-(4-Fluoro-3-(2-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[1,5-a]pyrazine-7-carbonyl)benzyl)phthalazin-1(2H)-one

Precaución: for Research Use Only

Formula: C₂₂H₁₆F₄N₆O₂
MW: 472.4 g/mol
MDL Number: MFCD32899277
CAS Number: 1358715-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR