(S)-1-(4-Chlorophenyl)ethanamine hydrochloride ≥98%

Supplier: Ambeed
Danger

A1473823-1G A1473823-250MG
77592-762EA 22.55 USD
77592-762 77875-108
(S)-1-(4-Chlorophenyl)ethanamine hydrochloride ≥98%
(S)-1-(4-Chlorophenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁Cl₂N
MDL Number: MFCD12756979
CAS Number: 56782-68-4
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR