(S)-N-(1-(6-Aminopyridin-2-yl)ethyl)-5-(4-(trifluoromethyl)phenyl)-2-naphthamide ≥99%

Supplier: Ambeed
Warning

Synonyms: VT107

A1473528-1MG A1473528-5MG A1473528-10MG
77597-442EA 102.5 USD
77597-442 77597-444 77622-228
(S)-N-(1-(6-Aminopyridin-2-yl)ethyl)-5-(4-(trifluoromethyl)phenyl)-2-naphthamide ≥99%
(S)-N-(1-(6-Aminopyridin-2-yl)ethyl)-5-(4-(trifluoromethyl)phenyl)-2-naphthamide

Precaución: for Research Use Only

Formula: C₂₅H₂₀F₃N₃O
MW: 435.44 g/mol
CAS Number: 2417718-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR