1,3-Bis((S)-1-(naphthalen-1-yl)ethyl)-1H-imidazol-3-ium tetrafluoroborate ≥97%

Supplier: Ambeed
Danger

A1473476-1G A1473476-250MG A1473476-100MG
77597-434EA 387.45 USD
77597-434 77597-436 77622-224
1,3-Bis((S)-1-(naphthalen-1-yl)ethyl)-1H-imidazol-3-ium tetrafluoroborate ≥97%
1,3-Bis((S)-1-(naphthalen-1-yl)ethyl)-1H-imidazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₇H₂₅BF₄N₂
MW: 464.3054 g/mol
CAS Number: 569658-97-5
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR