(3S,5R)-5-(2,4-Difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-ylmethyl-p-toluenesulfonate ≥98%

Supplier: Ambeed
Danger

Synonyms: ((3R,5R)-5-((1H-1,2,4-Triazol-1-yl)methyl)-5-(2,4-difluorophenyl)tetrahydrofuran-3-yl)methyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid (3S,5R)-5-(2,4-Difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-ylmethyl Ester, ((3S,5R)-5-((1H-1,2,4-Triazol-1-yl)methyl)-5-(2,4-difluorophenyl)tetrahydrofuran-3-yl)methyl 4-methylbenzenesulphonate, Posaconazole Impurity 29

A1473207-1G A1473207-5G
77597-426EA 29.73 USD
77597-426 77624-258
(3S,5R)-5-(2,4-Difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-ylmethyl-p-toluenesulfonate ≥98%
(3S,5R)-5-(2,4-Difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-ylmethyl-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₂₁H₂₁F₂N₃O₄S
MW: 449.47 g/mol
MDL Number: MFCD13195567
CAS Number: 149809-43-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR