(R)-(4,4',6,6'-Tetramethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(bis(3,5-bis(trimethylsilyl)phenyl)phosphane) ≥98%

Supplier: Ambeed
Warning

A1473101-100MG A1473101-5G
77875-091EA 215 USD
77875-091 77875-092
(R)-(4,4',6,6'-Tetramethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(bis(3,5-bis(trimethylsilyl)phenyl)phosphane) ≥98%
(R)-(4,4',6,6'-Tetramethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(bis(3,5-bis(trimethylsilyl)phenyl)phosphane)
Formula: C₆₄H₁₀₀O₄P₂Si₈
CAS Number: 2829281-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR