(5-Chloro-2-(2-(methylamino)thiazol-4-yl)pyridin-4-yl)(4-(4-chlorobenzyl)piperazin-1-yl)methanone ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
WNK-IN-11
A147304-10MG
A147304-5MG
77748-027EA
45
USD
77748-027
77748-028
(5-Chloro-2-(2-(methylamino)thiazol-4-yl)pyridin-4-yl)(4-(4-chlorobenzyl)piperazin-1-yl)methanone ≥98%
(5-Chloro-2-(2-(methylamino)thiazol-4-yl)pyridin-4-yl)(4-(4-chlorobenzyl)piperazin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₂₁H₂₁Cl₂N₅OS MW: 462.4 g/mol |
MDL Number:
MFCD31744539 CAS Number: 2123489-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...