N-(7-((R)-3-((5-Chloro-4-((2-(isopropylsulfonyl)phenyl)amino)pyrimidin-2-yl)amino)piperidin-1-yl)heptyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide 98+%

Supplier: Ambeed
Warning

A1472916-100MG A1472916-50MG
77875-086EA 548 USD
77875-086 77875-087
N-(7-((R)-3-((5-Chloro-4-((2-(isopropylsulfonyl)phenyl)amino)pyrimidin-2-yl)amino)piperidin-1-yl)heptyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide 98+%
N-(7-((R)-3-((5-Chloro-4-((2-(isopropylsulfonyl)phenyl)amino)pyrimidin-2-yl)amino)piperidin-1-yl)heptyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide

Precaución: for Research Use Only

Formula: C₄₀H₄₉ClN₈O₈S
MDL Number: MFCD34186973
CAS Number: 2519823-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR