3-Carbamoyl-4-chlorophenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (3-Carbamoyl-4-chlorophenyl)boronic acid

A147289-100MG A147289-1G A147289-250MG
77871-718EA 18 USD
77871-718 77871-719 77871-720
3-Carbamoyl-4-chlorophenylboronic acid ≥98%
3-Carbamoyl-4-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClNO₃
MDL Number: MFCD07363796
CAS Number: 871332-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR