4-(1-Hydroxy-2-phenoxyethyl)-2-methoxyphenol ≥97%
Supplier: Ambeed
|
Warning
|
A1472762-100MG
A1472762-250MG
77875-083EA
61
USD
77875-083
77875-084
4-(1-Hydroxy-2-phenoxyethyl)-2-methoxyphenol ≥97%
4-(1-Hydroxy-2-phenoxyethyl)-2-methoxyphenol
Precaución: for Research Use Only
Formula:
C₁₅H₁₆O₄ MW: 260.2851 g/mol |
CAS Number:
82246-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...