N-(2-Chloro-3-methoxyphenyl)acetamide ≥97%

Supplier: Ambeed
Warning

A1471670-100MG A1471670-1G A1471670-250MG
77875-079EA 20 USD
77875-079 77875-080 77875-081
N-(2-Chloro-3-methoxyphenyl)acetamide ≥97%
N-(2-Chloro-3-methoxyphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MDL Number: MFCD10001496
CAS Number: 150637-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR