4-((5-Aminopentyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Thalidomide-NH-C5-NH2 HCl
A1471138-50MG
A1471138-100MG
77592-752EA
60.48
USD
77592-752
77624-252
4-((5-Aminopentyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%
4-((5-Aminopentyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride
Precaución: for Research Use Only
Formula:
C₁₈H₂₃ClN₄O₄ MW: 394.85 g/mol |
MDL Number:
MFCD31729299 CAS Number: 2375194-03-7 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...