tert-Butyl 2-(methylthio)-4-(((trifluoromethyl)sulfonyl)oxy)-5,8-dihydropyrido[3,4-d]pyrimidine-7(6H)-carboxylate ≥98%

Supplier: Ambeed
Danger

A1469995-100MG A1469995-1G A1469995-250MG
77875-060EA 46 USD
77875-060 77875-061 77875-062
tert-Butyl 2-(methylthio)-4-(((trifluoromethyl)sulfonyl)oxy)-5,8-dihydropyrido[3,4-d]pyrimidine-7(6H)-carboxylate ≥98%
tert-Butyl 2-(methylthio)-4-(((trifluoromethyl)sulfonyl)oxy)-5,8-dihydropyrido[3,4-d]pyrimidine-7(6H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₈F₃N₃O₅S₂
MDL Number: MFCD32671039
CAS Number: 338739-82-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR