4-((2-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)ethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: Pomalidomide-PEG3-C2-NH2 HCl

A1469654-100MG A1469654-250MG
77624-246EA 153.47 USD
77624-246 77624-248
4-((2-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)ethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%
4-((2-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)ethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₉ClN₄O₇
MW: 484.93 g/mol
MDL Number: MFCD32705248
CAS Number: 2446474-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR