4-((2-(2-Aminoethoxy)ethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%

Supplier: Ambeed
Danger

Synonyms: Thalidomide-NH-PEG1-NH2 HCl

A1469644-250MG A1469644-50MG A1469644-1G
77597-386EA 307.5 USD
77597-386 77597-388 77875-055
4-((2-(2-Aminoethoxy)ethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%
4-((2-(2-Aminoethoxy)ethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₁ClN₄O₅
MW: 396.8254 g/mol
MDL Number: MFCD32903278
CAS Number: 2154342-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR